Methyl-5,6-dihydroxypyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5,6-dihydroxypicolinate

115724-1G
102899-490EA 2692.31 USD
102899-490
Methyl-5,6-dihydroxypyridine-2-carboxylate ≥95%
Methyl-5,6-dihydroxypyridine-2-carboxylate

Hazard: Irritant

Formula: C₇H₇NO₄
MW: 169.14 g/mol
MDL Number: MFCD09838989
CAS Number: 1260883-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR