4-Methoxy-1H-indazole ≥95%

Supplier: Matrix Scientific

115717-1G
102899-476EA 1171.85 USD
102899-476
4-Methoxy-1H-indazole ≥95%
4-Methoxy-1H-indazole

Hazard: Irritant

Formula: C₈H₈N₂O
MW: 148.1619 g/mol
MDL Number: MFCD09261131
CAS Number: 351210-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR