Imidazole-2-amidine acetate ≥95%

Supplier: Matrix Scientific

115715-1G
102899-470EA 1896.92 USD
102899-470
Imidazole-2-amidine acetate ≥95%
Imidazole-2-amidine acetate

Hazard: Irritant

Formula: C₆H₁₀N₄O₂
MDL Number: MFCD20922762
CAS Number: 950685-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR