(S)-3-Hydroxymethylpyrrolidine hydrochloride ≥95%

Supplier: Matrix Scientific

115714-1G
102899-606EA 1787.69 USD
102899-606
(S)-3-Hydroxymethylpyrrolidine hydrochloride ≥95%
(S)-3-Hydroxymethylpyrrolidine hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClNO
MDL Number: MFCD09800471
CAS Number: 1316087-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR