3,8-Dichloro-1,5-naphthyridine ≥95%

Supplier: Matrix Scientific

115708-1G
102897-242EA 1978.46 USD
102897-242
3,8-Dichloro-1,5-naphthyridine ≥95%
3,8-Dichloro-1,5-naphthyridine

Hazard: Irritant

Formula: C₈H₄Cl₂N₂
MW: 199.04 g/mol
MDL Number: MFCD00034435
CAS Number: 28252-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR