2,4-Dichloro-6-methylthieno[2,3-d]pyrimidine ≥95%
Supplier: Matrix Scientific
115707-1G
102897-240EA
1429.23
USD
102897-240
2,4-Dichloro-6-methylthieno[2,3-d]pyrimidine ≥95%
2,4-Dichloro-6-methylthieno[2,3-d]pyrimidine
Hazard: Irritant
Formula:
C₇H₄Cl₂N₂S |
MDL Number:
MFCD15144570 CAS Number: 76872-23-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...