3-Cyano-3-methylazetidine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Methyl-3-azetidinecarbonitrile hydrochloride3-Methylazetidine-3-carbonitrile hydrochloride

115706-1G
102897-238EA 2307.69 USD
102897-238
3-Cyano-3-methylazetidine hydrochloride ≥95%
3-Cyano-3-methylazetidine hydrochloride

Hazard: Irritant

Formula: C₅H₉ClN₂
MW: 132.59 g/mol
MDL Number: MFCD11849039
CAS Number: 936850-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR