5-Chlorothiophene-3-methanol ≥95%

Supplier: Matrix Scientific

115703-1G
102897-232EA 1567.69 USD
102897-232
5-Chlorothiophene-3-methanol ≥95%
5-Chlorothiophene-3-methanol

Hazard: Irritant

Formula: C₅H₅ClOS
MDL Number: MFCD13659394
CAS Number: 73919-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR