4-Chlorothiophene-2-methanol ≥95%

Supplier: Matrix Scientific

115702-1G
102897-230EA 684.62 USD
102897-230
4-Chlorothiophene-2-methanol ≥95%
4-Chlorothiophene-2-methanol

Hazard: Irritant

Formula: C₅H₅ClOS
MDL Number: MFCD17016116
CAS Number: 233280-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR