7-Bromo-2,4-dichloropyrrolo[2,1-f][1,2,4]triazine ≥92%

Supplier: Matrix Scientific

115691-1G
102897-224EA 3076.92 USD
102897-224
7-Bromo-2,4-dichloropyrrolo[2,1-f][1,2,4]triazine ≥92%
7-Bromo-2,4-dichloropyrrolo[2,1-f][1,2,4]triazine

Hazard: Irritant

Formula: C₆H₂BrCl₂N₃
MDL Number: MFCD22415227
CAS Number: 1008112-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR