1-Boc-2-hydroxymethyl-4-methylpyrrolidine ≥95%

Supplier: Matrix Scientific

115689-1G
102897-220EA 3873.85 USD
102897-220
1-Boc-2-hydroxymethyl-4-methylpyrrolidine ≥95%
1-Boc-2-hydroxymethyl-4-methylpyrrolidine

Hazard: Irritant

Formula: C₁₁H₂₁NO₃
MDL Number: MFCD22421683
CAS Number: 540501-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR