3-Boc-8-hydroxy-3-azabicyclo[3.2.1]octane ≥95%

Supplier: Matrix Scientific

115688-1G
102897-218EA 2307.69 USD
102897-218
3-Boc-8-hydroxy-3-azabicyclo[3.2.1]octane ≥95%
3-Boc-8-hydroxy-3-azabicyclo[3.2.1]octane

Hazard: Irritant

Formula: C₁₂H₂₁NO₃
MDL Number: MFCD22415201
CAS Number: 1330766-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR