2-Amino-7H-pyrrolo[2,3-d]pyrimidine ≥95%

Supplier: Matrix Scientific

Synonyms: 7H-Pyrrolo[2,3-d]pyrimidin-2-amine

115682-1G
102899-564EA 1567.69 USD
102899-564
2-Amino-7H-pyrrolo[2,3-d]pyrimidine ≥95%
2-Amino-7H-pyrrolo[2,3-d]pyrimidine

Hazard: Irritant

Formula: C₆H₆N₄
MW: 134.14 g/mol
MDL Number: MFCD12923656
CAS Number: 93366-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR