(3S,4S)-3-Amino-4-hydroxytetrahydropyran ≥95%

Supplier: Matrix Scientific

Synonyms: (3S,4S)-3-Aminotetrahydro-2H-pyran-4-ol

115679-1G
102897-208EA 1023.08 USD
102897-208
(3S,4S)-3-Amino-4-hydroxytetrahydropyran ≥95%
(3S,4S)-3-Amino-4-hydroxytetrahydropyran

Hazard: Irritant

Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
MDL Number: MFCD19215826
CAS Number: 1240390-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR