7-Amino-3-bromothieno[2,3-c]pyridine ≥95%

Supplier: Matrix Scientific

115676-1G
102897-194EA 2172.31 USD
102897-194
7-Amino-3-bromothieno[2,3-c]pyridine ≥95%
7-Amino-3-bromothieno[2,3-c]pyridine

Hazard: Irritant

Formula: C₇H₅BrN₂S
MW: 229.1 g/mol
MDL Number: MFCD22415204
CAS Number: 1326715-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR