4-Amino-5-bromo-2-methoxypyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-methoxypyridin-4-amine

115674-5G 115674-1G
102897-184EA 1183.08 USD
102897-184 102897-186
4-Amino-5-bromo-2-methoxypyridine ≥95%
4-Amino-5-bromo-2-methoxypyridine

Hazard: Irritant

Formula: C₆H₇BrN₂O
MW: 203.04 g/mol
MDL Number: MFCD12025502
CAS Number: 1232431-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR