6-Amino-3-bromoimidazo[1,2-a]pyridine ≥95%
Supplier: Matrix Scientific
115673-5G
115673-10G
115673-1G
102897-178EA
798.46
USD
102897-178
102897-180
102897-182
6-Amino-3-bromoimidazo[1,2-a]pyridine ≥95%
6-Amino-3-bromoimidazo[1,2-a]pyridine
Hazard: Irritant
Formula:
C₇H₆BrN₃ MW: 212.05 g/mol |
MDL Number:
MFCD11655113 CAS Number: 1099621-14-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...