Chloromethyl (trimethylsilyl)methylsulfide ≥95%

Supplier: Matrix Scientific

115668-1G
102897-150EA 604.62 USD
102897-150
Chloromethyl (trimethylsilyl)methylsulfide ≥95%
Chloromethyl (trimethylsilyl)methylsulfide

Hazard: Irritant

Formula: C₅H₁₃ClSSi
MDL Number: MFCD17013410
CAS Number: 105850-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR