7-Bromoimidazo[1,2-a]pyridine-2-carbaldehyde ≥95%
Supplier: Matrix Scientific
115659-1G
115659-5G
102899-542EA
3644.62
USD
102899-542
102899-540
7-Bromoimidazo[1,2-a]pyridine-2-carbaldehyde ≥95%
7-Bromoimidazo[1,2-a]pyridine-2-carbaldehyde
Hazard: Irritant
Formula:
C₈H₅BrN₂O |
MDL Number:
MFCD09995135 CAS Number: 1018828-16-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...