Methyl-2-chloroquinazoline-8-carboxylate ≥95%

Supplier: Matrix Scientific

115644-1G
102897-048EA 3160 USD
102897-048
Methyl-2-chloroquinazoline-8-carboxylate ≥95%
Methyl-2-chloroquinazoline-8-carboxylate

Hazard: Irritant

Formula: C₁₀H₇ClN₂O₂
MDL Number: MFCD15072116
CAS Number: 1217269-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR