2-(Hydrazinomethyl)pyrimidine ≥95%

Supplier: Matrix Scientific

115639-1G
102897-030EA 3350.77 USD
102897-030
2-(Hydrazinomethyl)pyrimidine ≥95%
2-(Hydrazinomethyl)pyrimidine

Hazard: Irritant

Formula: C₅H₈N₄
MDL Number: MFCD16848059
CAS Number: 1234616-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR