(S)-3-(Hydoxymethyl)-1-methylpyrrolidine ≥95%

Supplier: Matrix Scientific

115638-1G
102897-028EA 2172.31 USD
102897-028
(S)-3-(Hydoxymethyl)-1-methylpyrrolidine ≥95%
(S)-3-(Hydoxymethyl)-1-methylpyrrolidine

Hazard: Irritant

Formula: C₆H₁₃NO
MDL Number: MFCD17015955
CAS Number: 1210934-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR