tert-Butyl-3,3-difluoro-4-formyl-4-methylpiperidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
115627-250MG
115627-1G
102897-014EA
10080
USD
102897-014
102897-012
tert-Butyl-3,3-difluoro-4-formyl-4-methylpiperidine-1-carboxylate ≥95%
tert-Butyl-3,3-difluoro-4-formyl-4-methylpiperidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₂H₁₉F₂NO₃ MW: 263.29 g/mol |
CAS Number:
1400764-63-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...