5-Chloro-3-cyclopropylpyrazole ≥95%

Supplier: Matrix Scientific

115626-1G
102897-010EA 2363.08 USD
102897-010
5-Chloro-3-cyclopropylpyrazole ≥95%
5-Chloro-3-cyclopropylpyrazole

Hazard: Irritant

Formula: C₆H₇ClN₂
MDL Number: MFCD17214378
CAS Number: 1416437-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR