5-Bromoimidazo[1,2-a]pyridine-2-carbaldehyde ≥95%

Supplier: Matrix Scientific

115622-5G 115622-1G
102897-084EA 989.23 USD
102897-084 102897-086
5-Bromoimidazo[1,2-a]pyridine-2-carbaldehyde ≥95%
5-Bromoimidazo[1,2-a]pyridine-2-carbaldehyde

Hazard: Irritant

Formula: C₈H₅BrN₂O
MDL Number: MFCD11226918
CAS Number: 878197-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR