3-(Boc-aminomethyl)-3-methylazetidine oxalate ≥95%
Supplier: Matrix Scientific
115615-1G
115615-500MG
102897-058EA
1680
USD
102897-058
102896-984
3-(Boc-aminomethyl)-3-methylazetidine oxalate ≥95%
3-(Boc-aminomethyl)-3-methylazetidine oxalate
Hazard: Irritant
Formula:
C₁₂H₂₂N₂O₆ |
CAS Number:
1400764-23-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...