tert-Butyl-4-(aminomethyl)-3,3-difluoro-4-methylpiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115613-100MG 115613-500MG
102896-976EA 1647.69 USD
102896-976 102896-978
tert-Butyl-4-(aminomethyl)-3,3-difluoro-4-methylpiperidine-1-carboxylate ≥95%
tert-Butyl-4-(aminomethyl)-3,3-difluoro-4-methylpiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₂F₂N₂O₂
MW: 264.32 g/mol
MDL Number: MFCD19686629
CAS Number: 1334416-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR