5-(1-Aminoethyl)pyrimidine ≥95%

Supplier: Matrix Scientific

115612-1G
102899-506EA 1978.46 USD
102899-506
5-(1-Aminoethyl)pyrimidine ≥95%
5-(1-Aminoethyl)pyrimidine

Hazard: Irritant

Formula: C₆H₉N₃
MDL Number: MFCD10700207
CAS Number: 179323-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR