5-Amino-2,3-difluorophenol ≥95%

Supplier: Matrix Scientific

115611-100MG
102899-504EA 1373.85 USD
102899-504
5-Amino-2,3-difluorophenol ≥95%
5-Amino-2,3-difluorophenol

Hazard: Irritant

Formula: C₆H₅F₂NO
MDL Number: MFCD21601918
CAS Number: 1356339-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR