1-Piperidinecarboxylic acid-3-fluoro-4-[(phenylmethyl)amino]-(3S,4R)-1,1-dimethylethyl ester ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 3,4-cis-4-(benzylamino)-3-fluoropiperidine-1-carboxylate racemate
115605-500MG
115605-250MG
102897-002EA
2533.85
USD
102897-002
102897-000
1-Piperidinecarboxylic acid-3-fluoro-4-[(phenylmethyl)amino]-(3S,4R)-1,1-dimethylethyl ester ≥95%
1-Piperidinecarboxylic acid-3-fluoro-4-[(phenylmethyl)amino]-(3S,4R)-1,1-dimethylethyl ester
Hazard: Irritant
Formula:
C₁₇H₂₅FN₂O₂ MW: 308.4 g/mol |
MDL Number:
MFCD22421729 CAS Number: 907572-24-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...