7-Oxo-8-oxa-2,6-diazaspiro[4.5]decane ≥95%

Supplier: Matrix Scientific

115597-500MG
102896-960EA 4340 USD
102896-960
7-Oxo-8-oxa-2,6-diazaspiro[4.5]decane ≥95%
7-Oxo-8-oxa-2,6-diazaspiro[4.5]decane

Hazard: Irritant

Formula: C₇H₁₂N₂O₂
MW: 156.19 g/mol
MDL Number: MFCD22415225
CAS Number: 1389264-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR