2-Methoxyethylsulfonyl chloride ≥90%

Supplier: Matrix Scientific

Synonyms: 2-Methoxyethanesulfonyl chloride2-Methoxyethanesulphonyl chloride

115588-1G 115588-5G 115588-250MG
102896-920EA 275.38 USD
102896-920 102896-922 102896-924
2-Methoxyethylsulfonyl chloride ≥90%
2-Methoxyethylsulfonyl chloride

Hazard: Irritant

Formula: C₃H₇ClO₃S
MW: 158.6 g/mol
MDL Number: MFCD10032391
CAS Number: 51517-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR