1-(2-Ethoxyethynyl)cyclopentanol ≥95%

Supplier: Matrix Scientific

115582-250MG 115582-500MG
102896-894EA 2275.38 USD
102896-894 102896-892
1-(2-Ethoxyethynyl)cyclopentanol ≥95%
1-(2-Ethoxyethynyl)cyclopentanol

Hazard: Irritant

Formula: C₉H₁₄O₂
MDL Number: MFCD22421706
CAS Number: 100144-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR