α-(2-Ethoxyethynyl)cyclopentanemethanol ≥95%

Supplier: Matrix Scientific

115581-250MG 115581-1G
102896-888EA 989.23 USD
102896-888 102896-890
α-(2-Ethoxyethynyl)cyclopentanemethanol ≥95%
alpha-(2-Ethoxyethynyl)cyclopentanemethanol

Hazard: Irritant

Formula: C₁₀H₁₆O₂
MW: 168.24 g/mol
MDL Number: MFCD22421705
CAS Number: 1038404-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR