3-(3,3-Difluorocyclobutyl)-3-oxopropionitrile ≥95%
Supplier: Matrix Scientific
115580-1G
115580-500MG
102896-884EA
1593.85
USD
102896-884
102896-886
3-(3,3-Difluorocyclobutyl)-3-oxopropionitrile ≥95%
3-(3,3-Difluorocyclobutyl)-3-oxopropionitrile
Hazard: Irritant
Formula:
C₇H₇F₂NO |
MDL Number:
MFCD17016044 CAS Number: 1234616-26-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...