(R)-Pyrrolidine-3-carbonitrile hydrochloride ≥95%
Supplier: Matrix Scientific
115567-5G
115567-500MG
115567-1G
102896-850EA
3586.15
USD
102896-850
102896-848
102896-690
(R)-Pyrrolidine-3-carbonitrile hydrochloride ≥95%
(R)-Pyrrolidine-3-carbonitrile hydrochloride
Hazard: Irritant
Formula:
C₅H₉ClN₂ MW: 132.59 g/mol |
MDL Number:
MFCD11656839 CAS Number: 1153950-54-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...