(R)-3-Iodo-tetrahydrofuran ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-3-Iodotetrahydrofuran

115565-500MG 115565-5G
102896-684EA 2616.92 USD
102896-684 102896-680
(R)-3-Iodo-tetrahydrofuran ≥95%
(R)-3-Iodo-tetrahydrofuran

Hazard: Irritant

Formula: C₄H₇IO
MW: 198 g/mol
MDL Number: MFCD17015957
CAS Number: 873063-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR