O-4-Chlorobenzoylhydroxylamine ≥95%

Supplier: Matrix Scientific

115564-25G 115564-1G
102896-678EA 2375.38 USD
102896-678 102896-674
O-4-Chlorobenzoylhydroxylamine ≥95%
O-4-Chlorobenzoylhydroxylamine

Hazard: Irritant

Formula: C₇H₆ClNO₂
MDL Number: MFCD22370138
CAS Number: 872851-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR