1-Oxa-6-azaspiro[3.3]heptanetrifluoroacetate ≥90%
Supplier: Matrix Scientific
115555-1G
115555-100MG
115555-250MG
102896-824EA
2172.31
USD
102896-824
102896-826
102896-828
1-Oxa-6-azaspiro[3.3]heptanetrifluoroacetate ≥90%
1-Oxa-6-azaspiro[3.3]heptanetrifluoroacetate
Hazard: Irritant
Formula:
C₇H₁₀F₃NO₃ MW: 213.16 g/mol |
MDL Number:
MFCD22370115 CAS Number: 1303587-87-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...