[(1-Chloro-2,2-dimethylcyclopropyl)thio]benzene ≥95%

Supplier: Matrix Scientific

115542-100MG 115542-250MG
102896-768EA 1044.62 USD
102896-768 102896-770
[(1-Chloro-2,2-dimethylcyclopropyl)thio]benzene ≥95%
[(1-Chloro-2,2-dimethylcyclopropyl)thio]benzene

Hazard: Irritant

Formula: C₁₁H₁₃ClS
MDL Number: MFCD22370071
CAS Number: 5533-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR