tert-Butyl-3,7-diazabicyclo[3.3.1]nonane-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3,7-diazabicyclo[3.3.1]nonane-3-carboxylate

115527-250MG 115527-500MG 115527-1G
102896-646EA 2326.15 USD
102896-646 102896-648 102896-644
tert-Butyl-3,7-diazabicyclo[3.3.1]nonane-3-carboxylate ≥95%
tert-Butyl-3,7-diazabicyclo[3.3.1]nonane-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₂N₂O₂
MW: 226.32 g/mol
MDL Number: MFCD17016673
CAS Number: 227940-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR