5-(tert-Butoxycarbonyl)-3a,4,5,6,7,7a-hexahydroisoxazolo[4,5-c]pyridine-3-carboxylic acid ≥95%
Supplier: Matrix Scientific
115519-250MG
115519-1G
115519-500MG
102896-612EA
2275.38
USD
102896-612
102896-614
102896-610
5-(tert-Butoxycarbonyl)-3a,4,5,6,7,7a-hexahydroisoxazolo[4,5-c]pyridine-3-carboxylic acid ≥95%
5-(tert-Butoxycarbonyl)-3a,4,5,6,7,7a-hexahydroisoxazolo[4,5-c]pyridine-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₈N₂O₅ |
MDL Number:
MFCD12198802 CAS Number: 1251012-71-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...