7-Azabicyclo[2.2.1]heptane-2-carboxylic acid ethyl ester hydrochloride ≥95%
Supplier: Matrix Scientific
115509-250MG
115509-500MG
115509-1G
102896-554EA
3683.08
USD
102896-554
102896-556
102896-552
7-Azabicyclo[2.2.1]heptane-2-carboxylic acid ethyl ester hydrochloride ≥95%
7-Azabicyclo[2.2.1]heptane-2-carboxylic acid ethyl ester hydrochloride
Hazard: Irritant
Formula:
C₉H₁₆ClNO₂ |
CAS Number:
1379448-58-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...