2-(3-(Aminomethyl)oxetan-3-yl)ethanol ≥95%
Supplier: Matrix Scientific
115508-250MG
115508-500MG
115508-100MG
102896-546EA
1044.62
USD
102896-546
102896-548
102896-550
2-(3-(Aminomethyl)oxetan-3-yl)ethanol ≥95%
2-(3-(Aminomethyl)oxetan-3-yl)ethanol
Hazard: Irritant
Formula:
C₆H₁₃NO₂ |
MDL Number:
MFCD22370034 CAS Number: 1045709-42-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...