4-(Trifluoroacetyl)piperidine hydrochloride ≥95%
Supplier: Matrix Scientific
115507-500MG
115507-1G
115507-5G
102896-542EA
2326.15
USD
102896-542
102896-544
102896-540
4-(Trifluoroacetyl)piperidine hydrochloride ≥95%
4-(Trifluoroacetyl)piperidine hydrochloride
Hazard: Irritant
Formula:
C₇H₁₁ClF₃NO |
MDL Number:
MFCD22041637 CAS Number: 1372103-92-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...