tert-Butyl-5-(imino(methoxy)methyl)-4-(trifluoro-methyl)-3,4-dihydropyridine-1(2H)-carboxylate ≥95%

Supplier: Matrix Scientific

115506-100MG
102896-538EA 1373.85 USD
102896-538
tert-Butyl-5-(imino(methoxy)methyl)-4-(trifluoro-methyl)-3,4-dihydropyridine-1(2H)-carboxylate ≥95%
tert-Butyl-5-(imino(methoxy)methyl)-4-(trifluoro-methyl)-3,4-dihydropyridine-1(2H)-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₉F₃N₂O₃
MW: 308.3 g/mol
MDL Number: MFCD22199365
CAS Number: 1373503-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR