tert-Butyl-3-fluoro-4-(hydroxymethyl)-5,6-dihydropyridine-1(2H)-carboxylate ≥80%
Supplier: Matrix Scientific
115500-250MG
115500-500MG
115500-1G
102896-384EA
4135.38
USD
102896-384
102896-386
102896-382
tert-Butyl-3-fluoro-4-(hydroxymethyl)-5,6-dihydropyridine-1(2H)-carboxylate ≥80%
tert-Butyl-3-fluoro-4-(hydroxymethyl)-5,6-dihydropyridine-1(2H)-carboxylate
Hazard: Irritant
Formula:
C₁₁H₁₈FNO₃ MW: 231.27 g/mol |
MDL Number:
MFCD22199426 CAS Number: 1237526-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...