tert-Butyl-4-(2-ethoxy-2-oxoethyl)-3,3-difluoropiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-(2-ethoxy-2-oxoethyl)-3,3-difluoropiperidine-1-carboxylate

115496-100MG
102899-416EA 1373.85 USD
102899-416
tert-Butyl-4-(2-ethoxy-2-oxoethyl)-3,3-difluoropiperidine-1-carboxylate ≥95%
tert-Butyl-4-(2-ethoxy-2-oxoethyl)-3,3-difluoropiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₂₃F₂NO₄
MW: 307.34 g/mol
MDL Number: MFCD19686697
CAS Number: 1334412-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR