2-(1-(tert-Butoxycarbonyl)-3-fluoropiperidin-4-yl)acetic acid ≥95%

Supplier: Matrix Scientific

115481-100MG
102896-460EA 1867.69 USD
102896-460
2-(1-(tert-Butoxycarbonyl)-3-fluoropiperidin-4-yl)acetic acid ≥95%
2-(1-(tert-Butoxycarbonyl)-3-fluoropiperidin-4-yl)acetic acid

Hazard: Irritant

Formula: C₁₂H₂₀FNO₄
MDL Number: MFCD22199382
CAS Number: 1373502-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR