1-Benzyl-3-(hydroxymethyl)piperidin-4-ol ≥95%

Supplier: Matrix Scientific

115456-500MG
102896-298EA 660 USD
102896-298
1-Benzyl-3-(hydroxymethyl)piperidin-4-ol ≥95%
1-Benzyl-3-(hydroxymethyl)piperidin-4-ol

Hazard: Irritant

Formula: C₁₃H₁₉NO₂
MDL Number: MFCD03940529
CAS Number: 886-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR